The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143836.2 | NP_001137308.1 | 9 - 10 | 1168 | 64 |
NM_001143837.1 | NP_001137309.1 | 12 - 13 | 1313 | 64 | |
NM_001291926.1 | NP_001278855.1 | 9 - 10 | 1164 | 64 | |
NM_001291927.1 | NP_001278856.1 | 9 - 10 | 918 | 64 | |
NM_001291929.1 | NP_001278858.1 | 8 - 9 | 831 | 64 | |
NM_001300995.1 | NP_001287924.1 | 9 - 10 | 891 | 64 | |
NM_016931.4 | NP_058627.1 | 9 - 10 | 1168 | 64 | |
XM_006718849.3 | XP_006718912.1 | 9 - 10 | 1096 | 64 | |
XM_011542857.2 | XP_011541159.1 | 9 - 10 | 968 | 64 | |
XM_017017841.1 | XP_016873330.1 | 8 - 9 | 1015 | 64 | |
XM_017017842.1 | XP_016873331.1 | 5 - 6 | 467 | 64 | |
XM_017017843.1 | XP_016873332.1 | 9 - 10 | 1096 | 64 | |
XM_017017844.1 | XP_016873333.1 | 10 - 11 | 1154 | 64 | |
XM_017017845.1 | XP_016873334.1 | 6 - 7 | 570 | 64 | |
GenBank mRNA | AB041035.1 | - | 9 - 10 | 1088 | 64 |
AF254621.1 | - | 9 - 10 | 935 | 64 | |
AF261943.1 | - | 9 - 10 | 849 | 64 | |
AJ704725.1 | - | 9 - 10 | 845 | 64 | |
AJ704726.1 | - | 9 - 10 | 849 | 64 | |
AK057189.1 | - | 12 - 13 | 1313 | 64 | |
AK291830.1 | - | 9 - 10 | 931 | 64 | |
AK298328.1 | - | 9 - 10 | 918 | 64 | |
AK298357.1 | - | 8 - 9 | 822 | 64 | |
AK298376.1 | - | 5 - 6 | 609 | 64 | |
AK311381.1 | - | 8 - 9 | 831 | 64 | |
AY288918.1 | - | 9 - 10 | 849 | 64 | |
BC040105.1 | - | 9 - 10 | 921 | 64 |