Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHE
Entrez Gene ID
126669
Gene Name
Src homology 2 domain containing E
Gene Aliases
-
Chromosome Location
Chr.1: 154469637 - 154502059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005244891.4 XP_005244948.1 5 - 6 1328 81

View Full Product Details