Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD9
Entrez Gene ID
122402
Gene Name
tudor domain containing 9
Gene Aliases
C14orf75, HIG-1, NET54
Chromosome Location
Chr.14: 103928343 - 104052667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153046.2 NP_694591.2 31 - 32 3651 74
XM_005267309.3 XP_005267366.1 30 - 31 3632 74
XM_006720019.3 XP_006720082.1 31 - 32 3788 74
XM_006720020.3 XP_006720083.1 30 - 31 3707 74
XM_011536397.2 XP_011534699.1 30 - 31 3683 74
XM_011536398.2 XP_011534700.1 31 - 32 3593 74
XM_011536400.2 XP_011534702.1 28 - 29 3527 74
XM_011536401.2 XP_011534703.1 31 - 32 3526 74
XM_011536402.2 XP_011534704.1 24 - 25 2624 74
XM_017020955.1 XP_016876444.1 29 - 30 3382 74
GenBank mRNA AK093483.1 - 4 - 5 581 74
AK097429.1 - 15 - 16 1721 74
AK097699.1 - 8 - 9 1123 74
AL833915.1 - 28 - 29 3041 74
BC128057.1 - 26 - 27 2789 74

View Full Product Details