The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164399.1 | NP_001157871.1 | 7 - 8 | 1002 | 139 |
XM_011537120.2 | XP_011535422.1 | 7 - 8 | 1439 | 139 | |
XM_011537122.2 | XP_011535424.1 | 7 - 8 | 1438 | 139 | |
XM_011537124.2 | XP_011535426.1 | 7 - 8 | 1438 | 139 | |
XM_011537125.2 | XP_011535427.1 | 7 - 8 | 1443 | 139 | |
XM_011537130.2 | XP_011535432.1 | 7 - 8 | 1441 | 139 | |
XM_017021621.1 | XP_016877110.1 | 7 - 8 | 1441 | 139 |