Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTOG
Entrez Gene ID
340990
Gene Name
otogelin
Gene Aliases
DFNB18B, MLEMP, OTGN
Chromosome Location
Chr.11: 17547373 - 17645944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277269.1 NP_001264198.1 49 - 50 8040 71
NM_001292063.1 NP_001278992.1 50 - 51 8004 71
GenBank mRNA DV080398.1 - 2 - 3 71

View Full Product Details