Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTOG
Entrez Gene ID
340990
Gene Name
otogelin
Gene Aliases
DFNB18B, MLEMP, OTGN
Chromosome Location
Chr.11: 17547373 - 17645944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277269.1 NP_001264198.1 4 - 5 428 79
NM_001292063.1 NP_001278992.1 5 - 6 392 79
GenBank mRNA EG328176.1 - 1 - 2 79

View Full Product Details