The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277269.1 | NP_001264198.1 | 5 - 6 | 582 | 63 |
NM_001292063.1 | NP_001278992.1 | 6 - 7 | 546 | 63 | |
GenBank mRNA | DV080384.1 | - | 3 - 4 | 63 |