Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOG
Entrez Gene ID
340990
Gene Name
otogelin
Gene Aliases
DFNB18B, MLEMP, OTGN
Chromosome Location
Chr.11: 17547373 - 17645944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277269.1 NP_001264198.1 7 - 8 898 60
NM_001292063.1 NP_001278992.1 8 - 9 862 60

View Full Product Details