Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP27C1
Entrez Gene ID
339761
Gene Name
cytochrome P450 family 27 subfamily C member 1
Gene Aliases
-
Chromosome Location
Chr.2: 127183832 - 127220096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001665.3 NP_001001665.3 5 - 6 850 90
XM_017003960.1 XP_016859449.1 6 - 7 1040 90
XM_017003961.1 XP_016859450.1 4 - 5 741 90
GenBank mRNA AK131190.1 - 5 - 6 831 90
BC039307.1 - 5 - 6 850 90

View Full Product Details