The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002147.2 | 15 - 16 | 2179 | 63 | |
GenBank mRNA | AK025690.1 | - | 15 - 16 | 2179 | 63 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | karyopherin subunit alpha 7 | KPNA7 | IPOA8 | -78.3kb | Anti-sense | Chr. - 99145467 - 99219417 on Build GRCh38 |
Downstream | actin related protein 2/3 complex subunit 1A | ARPC1A | Arc40, HEL-68, HEL-S-307, SOP2Hs, SOP2L | 27.9kb | Sense | Chr. - 99325873 - 99366262 on Build GRCh38 |