Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNAT3
Entrez Gene ID
346562
Gene Name
G protein subunit alpha transducin 3
Gene Aliases
GDCA
Chromosome Location
Chr.7: 80457291 - 80512503 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102386.2 NP_001095856.1 7 - 8 961 78
GenBank mRNA BC147016.1 - 7 - 8 961 78
BC147017.1 - 7 - 8 961 78

View Full Product Details