The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080508.2 | NP_001073977.1 | 1 - 2 | 351 | 136 |
XM_006715602.3 | XP_006715665.2 | 1 - 2 | 290 | 136 | |
XM_011536247.2 | XP_011534549.2 | 1 - 2 | 290 | 136 | |
XM_011536248.2 | XP_011534550.2 | 1 - 2 | 290 | 136 | |
XM_011536249.2 | XP_011534551.2 | 1 - 2 | 290 | 136 | |
XM_017011473.1 | XP_016866962.1 | 1 - 2 | 290 | 136 | |
GenBank mRNA | BC132715.1 | - | 1 - 2 | 351 | 136 |
BC157841.1 | - | 1 - 2 | 344 | 136 |