The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080508.2 | NP_001073977.1 | 4 - 5 | 822 | 67 |
XM_006715602.3 | XP_006715665.2 | 4 - 5 | 761 | 67 | |
XM_011536247.2 | XP_011534549.2 | 4 - 5 | 761 | 67 | |
XM_011536248.2 | XP_011534550.2 | 4 - 5 | 761 | 67 | |
XM_011536249.2 | XP_011534551.2 | 4 - 5 | 761 | 67 | |
XM_017011472.1 | XP_016866961.1 | 4 - 5 | 1208 | 67 | |
XM_017011473.1 | XP_016866962.1 | 4 - 5 | 761 | 67 | |
GenBank mRNA | AJ010278.1 | - | 3 - 4 | 326 | 67 |
AK310179.1 | - | 4 - 5 | 500 | 67 | |
BC040697.2 | - | 4 - 5 | 517 | 67 | |
BC132715.1 | - | 4 - 5 | 822 | 67 | |
BC157841.1 | - | 4 - 5 | 815 | 67 |