The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207437.3 | NP_997320.2 | 22 - 23 | 3671 | 72 |
XM_005253555.3 | XP_005253612.1 | 23 - 24 | 4334 | 72 | |
XM_011538014.2 | XP_011536316.1 | 23 - 24 | 4334 | 72 | |
XM_011538015.2 | XP_011536317.1 | 23 - 24 | 4334 | 72 | |
XM_011538016.2 | XP_011536318.1 | 23 - 24 | 4334 | 72 | |
XM_011538017.2 | XP_011536319.1 | 23 - 24 | 4334 | 72 | |
XM_011538019.2 | XP_011536321.1 | 5 - 6 | 933 | 72 | |
XM_011538020.2 | XP_011536322.1 | 23 - 24 | 4342 | 72 | |
XM_017018960.1 | XP_016874449.1 | 23 - 24 | 4334 | 72 | |
XM_017018961.1 | XP_016874450.1 | 22 - 23 | 4025 | 72 | |
XM_017018962.1 | XP_016874451.1 | 5 - 6 | 855 | 72 | |
GenBank mRNA | AB290168.1 | - | 2 - 3 | 190 | 72 |
DV080370.1 | - | 2 - 3 | 72 |