Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
Chromosome Location
Chr.12: 123761995 - 123935720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207437.3 NP_997320.2 5 - 6 467 140
XM_005253555.3 XP_005253612.1 5 - 6 959 140
XM_011538014.2 XP_011536316.1 5 - 6 959 140
XM_011538015.2 XP_011536317.1 5 - 6 959 140
XM_011538016.2 XP_011536318.1 5 - 6 959 140
XM_011538017.2 XP_011536319.1 5 - 6 959 140
XM_011538020.2 XP_011536322.1 5 - 6 967 140
XM_017018960.1 XP_016874449.1 5 - 6 959 140
XM_017018961.1 XP_016874450.1 5 - 6 959 140
GenBank mRNA DV080260.1 - 2 - 3 140

View Full Product Details