The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080534.1 | NP_001074003.1 | 10 - 11 | 3984 | 72 |
XM_005254394.4 | XP_005254451.1 | 11 - 12 | 4935 | 72 | |
XM_017022220.1 | XP_016877709.1 | 15 - 16 | 4855 | 72 | |
XM_017022221.1 | XP_016877710.1 | 14 - 15 | 4735 | 72 | |
XM_017022222.1 | XP_016877711.1 | 13 - 14 | 4633 | 72 | |
XM_017022223.1 | XP_016877712.1 | 11 - 12 | 10161 | 72 | |
XM_017022224.1 | XP_016877713.1 | 10 - 11 | 4861 | 72 | |
XM_017022225.1 | XP_016877714.1 | 8 - 9 | 2286 | 72 | |
GenBank mRNA | AK091491.1 | - | 7 - 8 | 960 | 72 |
BC039494.1 | - | 3 - 4 | 559 | 72 |