The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040709.1 | NP_001035799.1 | 3 - 4 | 623 | 121 |
XM_011541283.2 | XP_011539585.1 | 3 - 4 | 623 | 121 | |
XM_011541284.2 | XP_011539586.1 | 3 - 4 | 623 | 121 | |
XM_011541285.1 | XP_011539587.1 | 3 - 4 | 623 | 121 | |
GenBank mRNA | AB158472.1 | - | 3 - 4 | 257 | 121 |
AI983207.1 | - | 3 - 4 | 476 | 121 | |
AK131459.1 | - | 3 - 4 | 623 | 121 | |
AK289513.1 | - | 3 - 4 | 557 | 121 | |
AK293082.1 | - | 3 - 4 | 499 | 121 | |
AK302566.1 | - | 3 - 4 | 543 | 121 |