Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD18B
Entrez Gene ID
441459
Gene Name
ankyrin repeat domain 18B
Gene Aliases
bA255A11.3, bA255A11.5
Chromosome Location
Chr.9: 33524403 - 33578755 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011517881.2 XP_011516183.1 6 - 7 897 157

View Full Product Details