Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NWD1
Entrez Gene ID
284434
Gene Name
NACHT and WD repeat domain containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 16719526 - 16817963 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007525.3 NP_001007526.3 6 - 7 2192 72
NM_001290355.1 NP_001277284.1 7 - 8 2844 72
XM_011527929.1 XP_011526231.1 7 - 8 2376 72
XM_011527930.2 XP_011526232.1 7 - 8 2862 72
XM_011527931.2 XP_011526233.1 6 - 7 2642 72
XM_011527934.1 XP_011526236.1 2 - 3 581 72
XM_011527935.1 XP_011526237.1 7 - 8 2376 72
XM_017026669.1 XP_016882158.1 8 - 9 2929 72
GenBank mRNA BC117698.1 - 5 - 6 1840 72
BX648940.1 - 8 - 9 2844 72
CR749794.1 - 6 - 7 2188 72

View Full Product Details