Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDZD7
Entrez Gene ID
79955
Gene Name
PDZ domain containing 7
Gene Aliases
PDZK7
Chromosome Location
Chr.10: 101007683 - 101031157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195263.1 NP_001182192.1 16 - 17 2972 57
XM_011540177.2 XP_011538479.1 17 - 18 3132 57
XM_011540178.2 XP_011538480.1 16 - 17 2944 57
GenBank mRNA AK024422.1 - 12 - 13 3269 57
FJ617449.1 - 17 - 18 2850 57

View Full Product Details