Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NWD1
Entrez Gene ID
284434
Gene Name
NACHT and WD repeat domain containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 16719526 - 16817963 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007525.3 NP_001007526.3 13 - 14 3363 78
NM_001290355.1 NP_001277284.1 14 - 15 4015 78
XM_011527929.1 XP_011526231.1 14 - 15 3547 78
XM_011527930.2 XP_011526232.1 14 - 15 4033 78
XM_011527931.2 XP_011526233.1 13 - 14 3813 78
XM_011527934.1 XP_011526236.1 9 - 10 1752 78
XM_011527935.1 XP_011526237.1 14 - 15 3547 78
XM_017026669.1 XP_016882158.1 15 - 16 4100 78
GenBank mRNA BC117698.1 - 12 - 13 3011 78
BX648940.1 - 15 - 16 4015 78
CR749794.1 - 13 - 14 3359 78

View Full Product Details