The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033517.1 | NP_277052.1 | 17 - 18 | 2111 | 71 |
GenBank mRNA | AB569469.1 | - | 17 - 18 | 2111 | 71 |