Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
Chromosome Location
Chr.3: 57293699 - 57545203 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291661.1 NP_001278590.1 42 - 43 6946 118
XM_011533466.1 XP_011531768.1 42 - 43 6931 118
XM_011533467.1 XP_011531769.1 42 - 43 6898 118
XM_011533468.1 XP_011531770.1 42 - 43 6931 118
XM_011533469.1 XP_011531771.1 42 - 43 6931 118
XM_011533471.2 XP_011531773.1 42 - 43 6931 118
XM_017005860.1 XP_016861349.1 42 - 43 6931 118
XM_017005861.1 XP_016861350.1 41 - 42 7614 118
XM_017005862.1 XP_016861351.1 21 - 22 3506 118

View Full Product Details