Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOL1
Entrez Gene ID
131149
Gene Name
otolin 1
Gene Aliases
C1QTNF15
Chromosome Location
Chr.3: 161496808 - 161503942 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080440.1 NP_001073909.1 1 - 2 364 71

View Full Product Details