The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033396.1 | 7 - 8 | 1296 | 81 | |
XM_017017774.1 | XP_016873263.1 | 6 - 7 | 1127 | 81 | |
XM_017017775.1 | XP_016873264.1 | 4 - 5 | 1103 | 81 | |
XM_017017776.1 | XP_016873265.1 | 4 - 5 | 1103 | 81 | |
GenBank mRNA | DN831898.1 | - | 2 - 3 | 81 |