The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010874.4 | NP_001010874.2 | 3 - 4 | 441 | 88 |
XM_005265663.2 | XP_005265720.1 | 3 - 4 | 441 | 88 | |
XM_005265664.2 | XP_005265721.1 | 3 - 4 | 441 | 88 | |
XM_005265665.3 | XP_005265722.1 | 3 - 4 | 441 | 88 | |
XM_017007959.1 | XP_016863448.1 | 3 - 4 | 441 | 88 | |
XM_017007960.1 | XP_016863449.1 | 3 - 4 | 441 | 88 | |
GenBank mRNA | AK290606.1 | - | 3 - 4 | 437 | 88 |
AL833108.1 | - | 3 - 4 | 442 | 88 | |
BC072463.1 | - | 3 - 4 | 441 | 88 | |
BX647590.1 | - | 3 - 4 | 378 | 88 |