Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
PDPK1 | NM_001261816.1 | 11 | 1443 | NP_001248745.1 |
NM_002613.4 | 11 | 1443 | NP_002604.1 | |
NM_031268.5 | 8 | 1062 | NP_112558.2 | |
XM_011522521.2 | 11 | 5131 | XP_011520823.1 | |
XM_011522523.2 | 8 | 4755 | XP_011520825.1 | |
XM_017023271.1 | 11 | 5136 | XP_016878760.1 | |
XM_017023272.1 | 11 | 1692 | XP_016878761.1 | |
XM_017023273.1 | 11 | 1401 | XP_016878762.1 | |
XM_017023274.1 | 12 | 1539 | XP_016878763.1 | |
XM_017023275.1 | 8 | 4761 | XP_016878764.1 | |
XM_017023276.1 | 8 | 1005 | XP_016878765.1 | |
XM_017023277.1 | 9 | 1103 | XP_016878766.1 | |
AB209835.1 | 11 | 1410 | BAD93072.1 | |
AF017995.1 | 11 | 1374 | AAC51825.1 | |
AK131060.1 | 11 | 1377 | ||
AK222581.1 | 11 | 1386 | BAD96301.1 | |
BC006339.2 | 8 | 1023 | AAH06339.2 | |
BC012103.1 | 11 | 1403 | AAH12103.1 | |
BC033494.1 | 8 | 975 | AAH33494.1 | |
CR536517.1 | 11 | 1294 | CAG38755.1 | |
Y15056.1 | 11 | 1373 | CAA75341.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv977895 | Chr16:2582644 - 2586873 on Build GRCh38 | Duplication | PDPK1 |
esv2422427 | Chr16:10001 - 2723164 on Build GRCh38 | Duplication | RHBDF1 MEIOB WDR24 NUBP2 CCNF LMF1 ABCA17P SNHG19 PDIA2 TPSB2 SNORA64 AXIN1 RPS2 MIR1225 UNKL MIR940 SNORA10 E4F1 LOC105371184 WASIR2 STUB1 RHBDL1 RPUSD1 DECR2 NME4 SOX8 LOC652276 RPL23AP5 LOC107987233 METTL26 HS3ST6 PGP MAPK8IP3 RNPS1 MIR662 FAM234A MIR4516 RPL3L AMDHD2 MRPL28 CACNA1H CAPN15 RNF151 MSRB1 HN1L SNRNP25 NME3 NTHL1 HBM BAIAP3 MPG LOC100134368 IFT140 KCTD5 SLC9A3R2 NHLRC4 RAB11FIP3 ERVK13-1 LUC7L TMEM204 MIR3677 ZNF598 POLR3K TBL3 RAB40C BRICD5 SYNGR3 CCDC78 LOC107984876 FAHD1 SNORA78 WFIKKN1 HAGH PRSS27 LINC00235 SSTR5-AS1 CLCN7 NDUFB10 C16orf91 TPSAB1 LOC105371045 TSC2 GFER ABCA3 MIR6859-4 LINC00254 TRAF7 MIR3178 TBC1D24 HBQ1 C16orf59 NPRL3 LOC729652 CEMP1 NTN3 HBZ LOC100287175 GNG13 FBXL16 PRSS29P MLST8 MSLN UBE2I LOC105371038 SSTR5 LOC106660606 LOC105371049 GNPTG HBA2 TPSD1 C1QTNF8 PTX4 PKD1 MIR3176 RAB26 PIGQ CASKIN1 IGFALS MIR6767 METRN TPSG1 RGS11 CRAMP1 ARHGDIG HAGHL ATP6V0C LOC101929280 ECI1 MIR4717 PDPK1 TELO2 LMF1-AS1 FLJ42627 MRPS34 MIR6768 LOC105371047 TMEM8A FAM173A MIR3177 TSR3 DDX11L10 MIR5587 MIR3180-5 CCDC154 DNASE1L2 LOC105371046 WDR90 CHTF18 NPW PRR35 RHOT2 JMJD8 EME2 NOXO1 PRR25 MIR6511B1 SPSB3 MCRIP2 NARFL SNORD60 SNHG9 HBA1 |
esv2760067 | Chr16:2486824 - 2820535 on Build GRCh38 | Gain+Loss | ATP6V0C MIR3178 TBC1D24 PDPK1 LOC652276 SRRM2-AS1 SRRM2 FLJ42627 PRSS27 CEMP1 PRSS33 KCTD5 PRSS41 PRSS21 TCEB2 AMDHD2 ERVK13-1 |
esv3637634 | Chr16:2586284 - 2610862 on Build GRCh38 | Gain | PDPK1 LOC652276 |
nsv833122 | Chr16:2501717 - 2663937 on Build GRCh38 | Loss | CEMP1 ATP6V0C MIR3178 TBC1D24 PDPK1 LOC652276 FLJ42627 AMDHD2 ERVK13-1 |
nsv1052260 | Chr16:2074546 - 2629655 on Build GRCh38 | Gain | ABCA3 TRAF7 ATP6V0C ECI1 MIR4717 MIR3178 TBC1D24 PDPK1 CCNF C16orf59 MIR6768 LOC729652 ABCA17P SNHG19 CEMP1 NTN3 MIR1225 MIR3180-5 MIR940 MLST8 E4F1 DNASE1L2 LOC106660606 LOC105371049 MIR3677 LOC652276 BRICD5 PKD1 MIR6511B1 PGP RAB26 RNPS1 SNORD60 CASKIN1 MIR6767 MIR4516 AMDHD2 TSC2 |
nsv833123 | Chr16:2561247 - 2764988 on Build GRCh38 | Gain+Loss | PRSS27 KCTD5 PDPK1 LOC652276 SRRM2-AS1 SRRM2 FLJ42627 ERVK13-1 |