Product Details

Assay Reference Genome
Location

Chr.9:127462339 on build GRCh38
Cytoband
9q33.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
90678
Gene Symbol
LRSAM1
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
Location
Chr.9:127451486-127503501 on build GRCh38
Assay Gene Location
Overlaps Exon 9 - Intron 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LRSAM1 NM_001005373.3 NP_001005373.1
NM_001005374.3 NP_001005374.1
NM_001190723.2 NP_001177652.1
NM_138361.5 NP_612370.3
XM_006717316.3 XP_006717379.1
XM_017015283.1 XP_016870772.1
XM_017015284.1 XP_016870773.1
AK056203.1 BAB71119.1
AK091589.1 BAC03703.1
AK226106.1
AY358830.1 AAQ89189.1
BC009239.2 AAH09239.1

View Full Product Details