The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321708.1 | NP_001308637.1 | 6 - 7 | 812 | 142 |
NM_001321709.1 | NP_001308638.1 | 7 - 8 | 590 | 142 | |
NM_001321710.1 | NP_001308639.1 | 6 - 7 | 778 | 142 | |
NM_004717.3 | NP_004708.1 | 6 - 7 | 812 | 142 | |
XM_011516682.2 | XP_011514984.2 | 6 - 7 | 809 | 142 | |
XM_017012780.1 | XP_016868269.1 | 6 - 7 | 810 | 142 | |
XM_017012781.1 | XP_016868270.1 | 6 - 7 | 809 | 142 | |
XM_017012782.1 | XP_016868271.1 | 6 - 7 | 809 | 142 | |
XM_017012783.1 | XP_016868272.1 | 6 - 7 | 809 | 142 | |
XM_017012785.1 | XP_016868274.1 | 4 - 5 | 997 | 142 | |
XM_017012786.1 | XP_016868275.1 | 6 - 7 | 497 | 142 | |
XM_017012787.1 | XP_016868276.1 | 5 - 6 | 362 | 142 | |
XM_017012788.1 | XP_016868277.1 | 7 - 8 | 543 | 142 | |
GenBank mRNA | AB209167.1 | - | 6 - 7 | 450 | 142 |