The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003302.2 | NP_003293.2 | 3 - 4 | 533 | 118 |
GenBank mRNA | AB628085.1 | - | 3 - 4 | 300 | 118 |
AB628086.1 | - | 3 - 4 | 313 | 118 | |
AB628087.1 | - | 2 - 3 | 235 | 118 | |
AJ001902.1 | - | 3 - 4 | 435 | 118 | |
AK056773.1 | - | 3 - 4 | 427 | 118 | |
AK291906.1 | - | 3 - 4 | 470 | 118 | |
AK309312.1 | - | 3 - 4 | 424 | 118 | |
BC002680.2 | - | 3 - 4 | 426 | 118 | |
BC004249.1 | - | 3 - 4 | 438 | 118 | |
BC004999.1 | - | 3 - 4 | 432 | 118 | |
BC021540.1 | - | 3 - 4 | 438 | 118 | |
BC028985.1 | - | 3 - 4 | 445 | 118 | |
BM800083.1 | - | 3 - 4 | 533 | 118 | |
CR749505.1 | - | 3 - 4 | 361 | 118 |