The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016269.4 | NP_057353.1 | 5 - 6 | 1826 | 69 |
XM_005263046.3 | XP_005263103.1 | 5 - 6 | 830 | 69 | |
XM_005263047.1 | XP_005263104.1 | 5 - 6 | 454 | 69 | |
XM_006714233.1 | XP_006714296.1 | 5 - 6 | 531 | 69 | |
GenBank mRNA | AF288571.1 | - | 5 - 6 | 1291 | 69 |
BC050632.2 | - | 5 - 6 | 728 | 69 |