Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H14
Entrez Gene ID
79882
Gene Name
zinc finger CCCH-type containing 14
Gene Aliases
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location
Chr.14: 88562909 - 88614720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160103.1 NP_001153575.1 11 - 12 1739 65
NM_001160104.1 NP_001153576.1 11 - 12 1739 65
NM_001326296.1 NP_001313225.1 10 - 11 1664 65
NM_001326297.1 NP_001313226.1 11 - 12 1920 65
NM_001326299.1 NP_001313228.1 10 - 11 1664 65
NM_001326300.1 NP_001313229.1 10 - 11 1624 65
NM_001326301.1 NP_001313230.1 10 - 11 1845 65
NM_001326302.1 NP_001313231.1 10 - 11 1624 65
NM_001326305.1 NP_001313234.1 9 - 10 1549 65
NM_001326307.1 NP_001313236.1 10 - 11 1664 65
NM_001326310.1 NP_001313239.1 11 - 12 1739 65
NM_001326312.1 NP_001313241.1 10 - 11 1664 65
NM_001326314.1 NP_001313243.1 9 - 10 1549 65
NM_001326315.1 NP_001313244.1 11 - 12 1920 65
NM_001326316.1 NP_001313245.1 11 - 12 1587 65
NM_024824.4 NP_079100.2 11 - 12 1739 65
XM_005268067.4 XP_005268124.1 11 - 12 1734 65
XM_005268068.4 XP_005268125.1 10 - 11 1659 65
XM_005268073.3 XP_005268130.1 3 - 4 961 65
XM_006720257.2 XP_006720320.1 2 - 3 886 65
XM_011537160.2 XP_011535462.1 11 - 12 1847 65
XM_011537161.2 XP_011535463.1 10 - 11 1578 65
XM_017021648.1 XP_016877137.1 10 - 11 1659 65
XM_017021649.1 XP_016877138.1 10 - 11 1659 65
XM_017021650.1 XP_016877139.1 10 - 11 1659 65
GenBank mRNA AK304275.1 - 11 - 12 1610 65
AY578060.1 - 11 - 12 1623 65
AY578063.1 - 4 - 5 461 65
BC011793.1 - 11 - 12 1605 65
BC023641.2 - 7 - 8 1245 65
BX248265.1 - 7 - 8 1261 65
BX248281.1 - 3 - 4 340 65
BX640716.1 - 11 - 12 1618 65
BX640876.1 - 1 - 2 121 65

View Full Product Details