Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H14
Entrez Gene ID
79882
Gene Name
zinc finger CCCH-type containing 14
Gene Aliases
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location
Chr.14: 88562909 - 88614720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160103.1 NP_001153575.1 2 - 3 310 90
NM_001160104.1 NP_001153576.1 2 - 3 310 90
NM_001326295.1 NP_001313224.1 2 - 3 310 90
NM_001326296.1 NP_001313225.1 2 - 3 310 90
NM_001326298.1 NP_001313227.1 2 - 3 310 90
NM_001326299.1 NP_001313228.1 2 - 3 310 90
NM_001326306.1 NP_001313235.1 2 - 3 310 90
NM_001326307.1 NP_001313236.1 2 - 3 310 90
NM_001326310.1 NP_001313239.1 2 - 3 310 90
NM_001326312.1 NP_001313241.1 2 - 3 310 90
NM_001326313.1 NP_001313242.1 2 - 3 310 90
NM_024824.4 NP_079100.2 2 - 3 310 90
NM_207660.3 NP_997543.1 2 - 3 310 90
NR_136936.1 2 - 3 310 90
XM_005268067.4 XP_005268124.1 2 - 3 305 90
XM_005268068.4 XP_005268125.1 2 - 3 305 90
XM_005268069.4 XP_005268126.1 2 - 3 305 90
XM_005268070.4 XP_005268127.1 2 - 3 305 90
XM_005268071.4 XP_005268128.1 2 - 3 290 90
XM_011537161.2 XP_011535463.1 2 - 3 305 90
XM_011537162.2 XP_011535464.1 2 - 3 305 90
XM_017021648.1 XP_016877137.1 2 - 3 305 90
XM_017021649.1 XP_016877138.1 2 - 3 305 90
XM_017021650.1 XP_016877139.1 2 - 3 305 90
GenBank mRNA AK021868.1 - 2 - 3 156 90
AK302136.1 - 2 - 3 236 90
AK303123.1 - 2 - 3 211 90
AK304275.1 - 2 - 3 181 90
AY578060.1 - 2 - 3 194 90
AY578061.1 - 2 - 3 249 90
BC011793.1 - 2 - 3 176 90
BX640716.1 - 2 - 3 189 90
DB452844.1 - 2 - 3 310 90

View Full Product Details