Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H14
Entrez Gene ID
79882
Gene Name
zinc finger CCCH-type containing 14
Gene Aliases
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location
Chr.14: 88562909 - 88614720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160103.1 NP_001153575.1 8 - 9 1352 133
NM_001160104.1 NP_001153576.1 8 - 9 1352 133
NM_001326295.1 NP_001313224.1 8 - 9 1352 133
NM_001326296.1 NP_001313225.1 8 - 9 1352 133
NM_001326297.1 NP_001313226.1 8 - 9 1533 133
NM_001326298.1 NP_001313227.1 8 - 9 1352 133
NM_001326299.1 NP_001313228.1 8 - 9 1352 133
NM_001326300.1 NP_001313229.1 7 - 8 1237 133
NM_001326301.1 NP_001313230.1 8 - 9 1533 133
NM_001326302.1 NP_001313231.1 7 - 8 1237 133
NM_001326303.1 NP_001313232.1 8 - 9 1533 133
NM_001326304.1 NP_001313233.1 7 - 8 1237 133
NM_001326305.1 NP_001313234.1 7 - 8 1237 133
NM_001326307.1 NP_001313236.1 8 - 9 1352 133
NM_001326308.1 NP_001313237.1 7 - 8 1237 133
NM_001326309.1 NP_001313238.1 7 - 8 1237 133
NM_001326310.1 NP_001313239.1 8 - 9 1352 133
NM_001326311.1 NP_001313240.1 7 - 8 1237 133
NM_001326312.1 NP_001313241.1 8 - 9 1352 133
NM_001326314.1 NP_001313243.1 7 - 8 1237 133
NM_001326315.1 NP_001313244.1 8 - 9 1533 133
NM_001326316.1 NP_001313245.1 8 - 9 1200 133
NM_024824.4 NP_079100.2 8 - 9 1352 133
NM_207660.3 NP_997543.1 8 - 9 1352 133
NM_207661.2 NP_997544.1 8 - 9 1200 133
NR_136936.1 8 - 9 1352 133
XM_005268067.4 XP_005268124.1 8 - 9 1347 133
XM_005268068.4 XP_005268125.1 8 - 9 1347 133
XM_005268069.4 XP_005268126.1 8 - 9 1347 133
XM_005268070.4 XP_005268127.1 8 - 9 1347 133
XM_005268071.4 XP_005268128.1 8 - 9 1332 133
XM_011537160.2 XP_011535462.1 8 - 9 1460 133
XM_011537162.2 XP_011535464.1 8 - 9 1347 133
XM_017021648.1 XP_016877137.1 8 - 9 1347 133
XM_017021649.1 XP_016877138.1 8 - 9 1347 133
XM_017021650.1 XP_016877139.1 8 - 9 1347 133
GenBank mRNA AK021868.1 - 8 - 9 1199 133
AK302136.1 - 8 - 9 1167 133
AK303123.1 - 9 - 10 1196 133
AK304275.1 - 8 - 9 1223 133
AL834215.1 - 8 - 9 1200 133
AY578060.1 - 8 - 9 1236 133
AY578061.1 - 8 - 9 1291 133
AY578062.1 - 8 - 9 1200 133
BC011793.1 - 8 - 9 1218 133
BC023641.2 - 4 - 5 858 133
BX248265.1 - 4 - 5 874 133
BX640716.1 - 8 - 9 1231 133

View Full Product Details