Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPN1
Entrez Gene ID
11321
Gene Name
GPN-loop GTPase 1
Gene Aliases
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1
Chromosome Location
Chr.2: 27628648 - 27650846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145047.1 NP_001138519.1 7 - 8 454 88
NM_001145048.1 NP_001138520.1 6 - 7 386 88
NM_001145049.1 NP_001138521.1 4 - 5 279 88
NM_007266.3 NP_009197.2 6 - 7 493 88
NR_026735.1 4 - 5 386 88
GenBank mRNA AB044661.1 - 6 - 7 454 88
AJ010842.1 - 6 - 7 383 88
AK298827.1 - 6 - 7 787 88
AK298866.1 - 6 - 7 493 88
AK302131.1 - 4 - 5 352 88
AK310340.1 - 7 - 8 444 88
AK311423.1 - 4 - 5 409 88
AK312992.1 - 6 - 7 465 88
AK316404.1 - 4 - 5 279 88
BC007451.2 - 6 - 7 437 88

View Full Product Details