Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPN1
Entrez Gene ID
11321
Gene Name
GPN-loop GTPase 1
Gene Aliases
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1
Chromosome Location
Chr.2: 27628648 - 27650846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145047.1 NP_001138519.1 10 - 11 741 105
NM_001145048.1 NP_001138520.1 9 - 10 673 105
NM_001145049.1 NP_001138521.1 7 - 8 566 105
NM_007266.3 NP_009197.2 9 - 10 780 105
NR_026735.1 7 - 8 673 105
GenBank mRNA AB044661.1 - 9 - 10 741 105
AJ010842.1 - 9 - 10 670 105
AK298827.1 - 9 - 10 1074 105
AK298866.1 - 9 - 10 780 105
AK302131.1 - 7 - 8 639 105
AK310340.1 - 10 - 11 731 105
AK311423.1 - 7 - 8 696 105
AK312992.1 - 9 - 10 752 105
AK316404.1 - 7 - 8 566 105
BC007451.2 - 9 - 10 724 105

View Full Product Details