The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018356.2 | NP_060826.2 | 5 - 6 | 999 | 100 |
NR_134298.1 | 5 - 6 | 775 | 100 | ||
XM_006714479.2 | XP_006714542.1 | 5 - 6 | 881 | 100 | |
XM_006714480.3 | XP_006714543.1 | 5 - 6 | 668 | 100 | |
XM_011514062.2 | XP_011512364.1 | 6 - 7 | 1077 | 100 | |
XM_017009607.1 | XP_016865096.1 | 6 - 7 | 1081 | 100 | |
XM_017009608.1 | XP_016865097.1 | 5 - 6 | 999 | 100 | |
XM_017009609.1 | XP_016865098.1 | 5 - 6 | 881 | 100 | |
XM_017009610.1 | XP_016865099.1 | 5 - 6 | 767 | 100 | |
XM_017009611.1 | XP_016865100.1 | 5 - 6 | 764 | 100 | |
XM_017009612.1 | XP_016865101.1 | 5 - 6 | 676 | 100 | |
XM_017009613.1 | XP_016865102.1 | 5 - 6 | 958 | 100 | |
XM_017009614.1 | XP_016865103.1 | 6 - 7 | 853 | 100 | |
XM_017009615.1 | XP_016865104.1 | 6 - 7 | 761 | 100 | |
XM_017009616.1 | XP_016865105.1 | 5 - 6 | 672 | 100 | |
GenBank mRNA | AK002055.1 | - | 5 - 6 | 986 | 100 |
BC021215.2 | - | 5 - 6 | 936 | 100 | |
BC032845.1 | - | 5 - 6 | 758 | 100 |