Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSH4
Entrez Gene ID
4438
Gene Name
mutS homolog 4
Gene Aliases
-
Chromosome Location
Chr.1: 75796871 - 75913238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002440.3 NP_002431.2 14 - 15 2026 72
GenBank mRNA AF104243.1 - 14 - 15 1922 72
BC033030.1 - 14 - 15 2016 72
U89293.1 - 14 - 15 1952 72

View Full Product Details