The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002440.3 | NP_002431.2 | 1 - 2 | 357 | 109 |
GenBank mRNA | AF104243.1 | - | 1 - 2 | 253 | 109 |
BC033030.1 | - | 1 - 2 | 347 | 109 | |
DB077677.1 | - | 1 - 2 | 357 | 109 | |
U89293.1 | - | 1 - 2 | 283 | 109 |