The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006610.3 | NP_006601.2 | 5 - 6 | 778 | 82 |
XM_017000097.1 | XP_016855586.1 | 5 - 6 | 778 | 82 | |
GenBank mRNA | AK290823.1 | - | 5 - 6 | 778 | 82 |
X98400.1 | - | 5 - 6 | 782 | 82 | |
Y09926.1 | - | 5 - 6 | 762 | 82 |