Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
Chromosome Location
Chr.18: 54357914 - 54382035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288980.1 NP_001275909.1 4 - 5 1375 75
NM_001288981.1 NP_001275910.1 4 - 5 1207 75
XM_005258201.1 XP_005258258.1 3 - 4 2076 75
XM_017025567.1 XP_016881056.1 3 - 4 1759 75
GenBank mRNA AB267906.1 - 3 - 4 2071 75
AB353076.1 - 4 - 5 1374 75
AB353077.1 - 4 - 5 1206 75
AK126503.1 - 2 - 3 813 75

View Full Product Details