The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005144.4 | NP_005135.2 | 12 - 13 | 3441 | 73 |
NM_018411.4 | NP_060881.2 | 12 - 13 | 3441 | 73 | |
XM_005273569.1 | XP_005273626.1 | 12 - 13 | 3445 | 73 | |
XM_006716367.1 | XP_006716430.1 | 12 - 13 | 3445 | 73 | |
GenBank mRNA | AF039196.3 | - | 12 - 13 | 4260 | 73 |
AJ277165.2 | - | 11 - 12 | 2816 | 73 | |
BC067128.1 | - | 12 - 13 | 2906 | 73 |