Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HR
Entrez Gene ID
55806
Gene Name
hair growth associated
Gene Aliases
ALUNC, AU, HSA277165, HYPT4, MUHH, MUHH1
Chromosome Location
Chr.8: 22114419 - 22131053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005144.4 NP_005135.2 16 - 17 3883 96
XM_005273569.1 XP_005273626.1 16 - 17 3887 96
GenBank mRNA AF039196.3 - 16 - 17 4702 96
AJ277165.2 - 15 - 16 3258 96
BC067128.1 - 16 - 17 3348 96

View Full Product Details