Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HR
Entrez Gene ID
55806
Gene Name
hair growth associated
Gene Aliases
ALUNC, AU, HSA277165, HYPT4, MUHH, MUHH1
Chromosome Location
Chr.8: 22114419 - 22131053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005144.4 NP_005135.2 2 - 3 1277 57
NM_018411.4 NP_060881.2 2 - 3 1277 57
XM_005273569.1 XP_005273626.1 2 - 3 1278 57
XM_006716367.1 XP_006716430.1 2 - 3 1278 57
GenBank mRNA AF039196.3 - 2 - 3 2096 57
AJ277165.2 - 1 - 2 652 57
BC067128.1 - 2 - 3 742 57

View Full Product Details