Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELP2
Entrez Gene ID
55250
Gene Name
elongator acetyltransferase complex subunit 2
Gene Aliases
SHINC-2, STATIP1, StIP
Chromosome Location
Chr.18: 36129874 - 36177931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242875.2 NP_001229804.1 8 - 9 908 69
NM_001242876.2 NP_001229805.1 7 - 8 830 69
NM_001242877.2 NP_001229806.1 6 - 7 635 69
NM_001242878.2 NP_001229807.1 6 - 7 635 69
NM_001324465.1 NP_001311394.1 7 - 8 713 69
NM_001324466.1 NP_001311395.1 7 - 8 830 69
NM_001324467.1 NP_001311396.1 7 - 8 713 69
NM_001324468.1 NP_001311397.1 6 - 7 634 69
NM_018255.3 NP_060725.1 7 - 8 713 69
NR_040110.2 7 - 8 713 69
NR_136898.1 7 - 8 713 69
NR_137173.1 7 - 8 713 69
GenBank mRNA AF332505.1 - 6 - 7 639 69
AF403223.1 - 7 - 8 690 69
AK001741.1 - 7 - 8 663 69
AK022709.1 - 6 - 7 639 69
AK222791.1 - 7 - 8 666 69
AK300201.1 - 7 - 8 784 69
AK302062.1 - 8 - 9 874 69
CR457269.1 - 7 - 8 653 69

View Full Product Details