The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282352.1 | NP_001269281.1 | 9 - 10 | 1156 | 64 |
NM_030957.3 | NP_112219.3 | 22 - 23 | 2967 | 64 | |
XM_006722917.3 | XP_006722980.1 | 21 - 22 | 2813 | 64 | |
XM_017027338.1 | XP_016882827.1 | 21 - 22 | 2823 | 64 | |
XM_017027339.1 | XP_016882828.1 | 12 - 13 | 1447 | 64 | |
XM_017027340.1 | XP_016882829.1 | 11 - 12 | 1307 | 64 | |
GenBank mRNA | AB209515.1 | - | 21 - 22 | 2803 | 64 |
AF163762.1 | - | 19 - 20 | 2583 | 64 | |
AK131414.1 | - | 9 - 10 | 1156 | 64 | |
BC029308.1 | - | 3 - 4 | 314 | 64 |