Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS10
Entrez Gene ID
81794
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Gene Aliases
ADAM-TS10, ADAMTS-10, WMS, WMS1
Chromosome Location
Chr.19: 8580240 - 8610735 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030957.3 NP_112219.3 7 - 8 1194 68
XM_006722917.3 XP_006722980.1 6 - 7 1040 68
XM_017027338.1 XP_016882827.1 6 - 7 1050 68
GenBank mRNA AB209515.1 - 6 - 7 1030 68
AF163762.1 - 4 - 5 810 68

View Full Product Details