The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003059.2 | NP_003050.2 | 8 - 9 | 1609 | 80 |
XM_006714675.3 | XP_006714738.1 | 7 - 8 | 1016 | 80 | |
XM_011543589.2 | XP_011541891.1 | 6 - 7 | 1401 | 80 | |
XM_017009776.1 | XP_016865265.1 | 8 - 9 | 1657 | 80 | |
GenBank mRNA | AB007448.1 | - | 8 - 9 | 1590 | 80 |
BC028313.1 | - | 8 - 9 | 1609 | 80 | |
Y09881.1 | - | 8 - 9 | 1602 | 80 |