Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARM1
Entrez Gene ID
25849
Gene Name
prostate androgen-regulated mucin-like protein 1
Gene Aliases
Cipar1, DKFZP564O0823, PARM-1, WSC4
Chromosome Location
Chr.4: 74933075 - 75050115 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015393.3 NP_056208.2 1 - 2 292 84
GenBank mRNA AK022311.1 - 1 - 2 251 84
AL080121.1 - 1 - 2 209 84
AY358777.1 - 1 - 2 237 84
CA390477.1 - 1 - 2 289 84

View Full Product Details