Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPFIA1
Entrez Gene ID
8500
Gene Name
PTPRF interacting protein alpha 1
Gene Aliases
LIP.1, LIP1, LIPRIN
Chromosome Location
Chr.11: 70270687 - 70384501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003626.3 NP_003617.1 25 - 26 3592 103
NM_177423.2 NP_803172.1 25 - 26 3592 103
NR_045286.1 26 - 27 3603 103
XM_006718716.3 XP_006718779.1 30 - 31 3917 103
XM_011545306.2 XP_011543608.1 30 - 31 3789 103
XM_011545307.2 XP_011543609.1 29 - 30 3890 103
XM_011545308.2 XP_011543610.1 29 - 30 3887 103
XM_011545309.2 XP_011543611.1 30 - 31 3917 103
XM_011545310.2 XP_011543612.1 29 - 30 3842 103
XM_011545311.2 XP_011543613.1 28 - 29 3830 103
XM_011545312.2 XP_011543614.1 28 - 29 3827 103
XM_011545313.2 XP_011543615.1 29 - 30 3800 103
XM_011545314.2 XP_011543616.1 28 - 29 3770 103
XM_011545315.2 XP_011543617.1 28 - 29 3725 103
XM_011545316.2 XP_011543618.1 27 - 28 3710 103
XM_011545317.2 XP_011543619.1 26 - 27 3680 103
XM_011545318.2 XP_011543620.1 15 - 16 2022 103
XM_011545319.1 XP_011543621.1 27 - 28 3698 103
XM_017018448.1 XP_016873937.1 27 - 28 3797 103
XM_017018449.1 XP_016873938.1 26 - 27 3635 103
GenBank mRNA AB209314.1 - 26 - 27 3530 103
AK123331.1 - - 2169 103
AK124621.1 - 5 - 6 2137 103
BC034046.1 - 25 - 26 3555 103
BX648037.1 - 24 - 25 2892 103
U22815.1 - 25 - 26 3606 103
U22816.1 - 25 - 26 3606 103

View Full Product Details